Retort rods, with M10 thread
12 mm, 1000 mm, M10
Katalognr. BOCH5153
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Galvanised steel.
- Rods for stand bases
- M10 thread
Technische Daten
- Ø:12 mm
- Gewinde:M10
- Länge:1000 mm
- Cat. No.:BOCH5153