AuftragsverfolgungKontakt
Mounting/reducer clips for tubes | Avantor
Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm), Tubes with different diameters
Katalognr. BOCH553
Anbieter:   BOCHEM
Mounting/reducer clips for tubes
Katalognr. BOCH553
Anbieter:  BOCHEM
Mounting/reducer clips for tubes
Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm), Tubes with different diameters
Preis
SFr. 192.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
Technische Daten
  • Für:
    Tubes with different diameters
  • Bezeichnung:
    Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm)
  • Seq:
    03