Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 15 mm), Tubes with different diameters
Catalog # BOCH551
Supplier: BOCHEM
Jump to:
Product Details & Documents
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
Specifications
- Für:Tubes with different diameters
- Bezeichnung:Mounting clip/reducer clamp (OD 26,9 mm, Ø 15 mm)
- Seq:05