Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm), Tubes with different diameters
Katalognr. BOCH554
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
Technische Daten
- Für:Tubes with different diameters
- Bezeichnung:Mounting clip/reducer clamp (OD 26,9 mm, Ø 12 mm)
- Seq:02