AuftragsverfolgungKontakt
Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 26,9 mm), Tubes with identical diameter
Katalognr. BOCH550
Anbieter:   BOCHEM
Mounting/reducer clips for tubes
Katalognr. BOCH550
Anbieter:  BOCHEM
Mounting/reducer clips for tubes
Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 26,9 mm), Tubes with identical diameter
Preis
SFr. 196.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
Technische Daten
  • Für:
    Tubes with identical diameter
  • Bezeichnung:
    Mounting clip/reducer clamp (OD 26,9 mm, Ø 26,9 mm)
  • Seq:
    01