Mounting/reducer clips for tubes
Mounting clip/reducer clamp (OD 26,9 mm, Ø 26,9 mm), Tubes with identical diameter
Katalognr. BOCH550
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Mounting clips/reducers made of stainless steel. Either for tubes with identical diameter or for tubes with different diameters.
Technische Daten
- Für:Tubes with identical diameter
- Bezeichnung:Mounting clip/reducer clamp (OD 26,9 mm, Ø 26,9 mm)
- Seq:01