Instrument trays
180×240×17 mm
Katalognr. BURK4230-3024
Anbieter: BURKLE
Direkt zu
Product Details & Documents
Melamine, white.
Tray without lid
Suitable for use with foodstuffs
Not autoclavable
Not suitable for high temperatures
Bestellinformationen: Lid available as accessory
Technische Daten
- B×T×H:180×240×17 mm
- Cat. No.:BURK4230-3024