Methanol ≥98,5%, TECHNICAL
Kunststoffflasche, 5 L
Anbieter: VWR Chemicals
Direkt zu
Product Details & Documents
Synonyme: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Nummer:67-56-1
- Formel:CH₃OH
- MDL-Nummer:MFCD00004595
- Molekulargewicht:32.04 g/mol
- Merck-Index:13,05984
- Siedepunkt:64,6 °C (1013 hPa)
- Schmelzender Platingehalt:–98 °C
- Dichte:0,7918 g/cm³ (20 °C)
- Blitz Pt:11 °C (geschlossener Tiegel)
- Lagertemperatur:Raumtemperatur
- EINECS:200-659-6
- UN:1230
- ADR:3 (6.1),II
- ERREICHEN:01-2119433307-44
Documents
Zertifikate
Suche nach Chargennummer10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951065)
Packaging Information
P20_VWRC20903.290_SPEC_DE.pdf P20_VWRC20903.368_SPEC_DE.pdf P20_VWRC20903.415_SPEC_DE.pdf P20_VWRC20903.461_SPEC_DE.pdf P20_VWRC20903.550_SPEC_DE.pdf
Product specification
Mehr
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Technische Daten
Ergebnisse des Spezifikationstests
- Gehalt:Min. 98,5 %
- Verdampfungsrückstand:Max. 0,015 %
Technische Daten
- Packung:Kunststoffflasche
- VE:5 L
- Cat. No.:20903.368
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



