AuftragsverfolgungKontakt
Test kits, digital titration method | Avantor
Test kits, digital titration method
Sulfuric acid digital titrator cartridge, 8,00 N, 1
Katalognr. HACH1439101
Anbieter:   Hach
Test kits, digital titration method
Katalognr. HACH1439101
Anbieter:  Hach
CAS Nummer:  
Test kits, digital titration method
Test kits, digital titration method
Sulfuric acid digital titrator cartridge, 8,00 N, 1
CAS Nummer:  
Preis
SFr. 67.80
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Achtung! Bitte beachten: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Sulfuric acid digital titrator cartridge, 8,00 N
Technische Daten
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..