Caps for bottles, open top, Microlink®, WHEATON®
Schraubverschluss, oben geschlossen, mit Polypropylen-/Silikon-/PTFE-Einlage, 38-430, Weiß, Labor-Packung
Catalog # WHEA240235-01
Supplier: DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Catalog # WHEA240235-01
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Specifications
- Gewinde:38-430
- Typ:Labor-Packung
- Farbe:Weiß
- Bezeichnung:Schraubverschluss, oben geschlossen, mit Polypropylen-/Silikon-/PTFE-Einlage
- Seq:14