Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Büchner-Trichter | Avantor

Büchner-Trichter

120 mm, 1000 ml, 1
Katalognr. 511-0778
Anbieter:   SciLabware
Büchner-Trichter
Katalognr. 511-0778
Anbieter:  SciLabware
Büchner-Trichter
Büchner-Trichter

120 mm, 1000 ml, 1

Preis
SFr. 509.00
Einloggen für Verfügbarkeit
1 * 2 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Technische Daten
  • Platte Ø:
    120 mm
  • Porosität:
    1
  • Kapazität:
    1000 ml
  • Seq:
    46
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.