Dissecting trays
310×200×45 mm
Katalognr. NICK625
Anbieter: NICKEL ELECTRO
Direkt zu
Product Details & Documents
Dissecting trays made of aluminium.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Technische Daten
- Seq:01
- B×T×H:310×200×45 mm