Retort rods, with M10 thread
13 mm, 1500 mm, M10
Katalognr. BOCH5175
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Stainless steel 18/10.
- Rods for stand bases
- M10 thread
Technische Daten
- Ø:13 mm
- Gewinde:M10
- Länge:1500 mm
- Seq:15
- Cat. No.:BOCH5175