Spatel, rund, gebogen
210 mm, 25×12 mm
Katalognr. BOCH3204
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.
- Excellent heat resistance, autoclavable
- Good resistance to chemicals
- Easy and quick cleaning
Dokumente
BOCH3204_TDS_EN
Technisches Datenblatt
Technische Daten
- Länge:210 mm
- Seq:08
- Löffelmaße:25×12 mm