Measuring Cylinders
250 ml, 2,0 ml, 315 mm, 2 ± ml
Catalog # 612-0430
Supplier: SciLabware
Jump to:
Product Details & Documents
PMP (TPX®), transparent, tall form, autoclavable, class B.
- Hexagonal base with non-slip, anti-suction feature, spout, excellent chemical resistance, no meniscus to confuse readings
- Recycling codes and batch code on each cylinder
- Autoclavable - Does not affect accuracy
Note: The product is suitable for lab washing machines, however, a temperature of below 60°C should be used to prevent damage.
Certifications: ISO 6706
Specifications
- Kapazität:250 ml
- Höhe:315 mm
- Teilung:2,0 ml
- Toleranz:2 ± ml