AuftragsverfolgungKontakt
Trolley, trays | Avantor
Trolley, trays
Trays trolley, without guard rails, two handles and two trays, knockdown construction, 500W×750L mm, 2
Katalognr. AGEN741
Anbieter:   Agencinox
Trolley, trays
Katalognr. AGEN741
Anbieter:  Agencinox
Trolley, trays
Trolley, trays
Trays trolley, without guard rails, two handles and two trays, knockdown construction, 500W×750L mm, 2
Preis
708.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.
  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models
Technische Daten
  • Bezeichnung:
    Trays trolley, without guard rails, two handles and two trays, knockdown construction, 500W×750L mm
  • Seq:
    03
  • Böden:
    2