Test Tubes, round bottom, with spout
25 ml, 180 mm, 18 mm
Katalognr. HECH42765025
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
AR® (soda glass).
- Fused white graduation
- Good chemical and physical properties
- Round bottom
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Technische Daten
- Ext.Ø:18 mm
- Kapazität:25 ml
- Länge:180 mm