AuftragsverfolgungKontakt
Hypodermic needles
Hypodermic needles
20, 63,5 mm
Katalognr. POPR4060
Anbieter:   CADENCE SCIENCE
Hypodermic needles
Katalognr. POPR4060
Anbieter:  CADENCE SCIENCE
Hypodermic needles
Hypodermic needles
20, 63,5 mm
Preis
SFr. 1’670.00
Einloggen für Verfügbarkeit
1 * 12 ST
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Technische Daten
  • Gauge:
    20
  • Länge:
    63,5 mm
  • Seq:
    14