Aceton ≥99,0% Ph. Eur.
Metalltrommel, 25 L
Anbieter: VWR Chemicals
Direkt zu
Product Details & Documents
Synonyme: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Nummer:67-64-1
- Formel:CH₃COCH₃
- MDL-Nummer:MFCD00008765
- Molekulargewicht:58.08 g/mol
- Merck-Index:13,00067
- Siedepunkt:56,2 °C (1013 hPa)
- Schmelzender Platingehalt:–95,4 °C
- Dichte:0,792 g/cm³ (20 °C)
- Blitz Pt:<–20 °C (geschlossener Tiegel)
- Lagertemperatur:Raumtemperatur
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- ERREICHEN:01-2119471330-49
Documents
Zertifikate
Suche nach Chargennummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
20165_ACETONE PH.EUR_AFLATOXIN_Rev1
AFLATOXIN
20165_ACETONE PH.EUR_BSE_Rev1
BSE/TSE Certificates
20165_ACETONE PH.EUR_LATEX Acetone Ph. Eur.
Additional Product Details
35008524.pdf P20_VWRC20165.460_SPEC_DE.pdf P20_VWRC20165.550_SPEC_DE.pdf spec_VWRC20165.298 spec_VWRC20165.298_de
Product specification
Mehr
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Technische Daten
Ergebnisse des Spezifikationstests
- Assay:Min. 99.0 %
- Appearance:Clear colourless liquid
- Identification B:Passes test
- Identification C:Passes test
- Appearance of solution:Passes test
- Acidity or alkalinity:Passes test
- Relative density:0.79 - 0.793
- Reducing substances:Passes test
- Methanol (V/V):Max. 0.05 %
- Propan-2-ol (V/V):Max. 0.05 %
- Benzene (V/V):Max. 2 ppm
- Any other impurity (V/V):Max. 0.05 %
- Matter insoluble in water:Passes test
- Residue on evaporation:Max. 50 ppm
- Water:Max. 3 g/l
- Residual solvents:Passes test
- Mesityl oxide (*):Max. 10 ppm
- Statement of original manufacturer:(*)
Technische Daten
- Packung:Metalltrommel
- VE:25 L
- Cat. No.:20165.460
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



