AuftragsverfolgungKontakt
EDTA solutions | Avantor
EDTA solutions
EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method, Marked dropping bottle, 50 mL
Katalognr. HACH2241926
Anbieter:   Hach
EDTA solutions
Katalognr. HACH2241926
Anbieter:  Hach
CAS Nummer:  
EDTA solutions
EDTA solutions
EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method, Marked dropping bottle, 50 mL
CAS Nummer:  
Preis
SFr. 38.80
Einloggen für Verfügbarkeit
1 * 50 mL
Product Details & Documents
Dokumente
Technische Daten
Technische Daten
  • Packung:
    Marked dropping bottle
  • VE:
    50 mL
  • Bezeichnung:
    EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method