AuftragsverfolgungKontakt
Test kits, digital titration method
Test kits, digital titration method
EDTA digital titrator cartridge, 0,716 M, 1
Katalognr. HACH2081801
Anbieter:   Hach
Test kits, digital titration method
Katalognr. HACH2081801
Anbieter:  Hach
CAS Nummer:  
Test kits, digital titration method
Test kits, digital titration method
EDTA digital titrator cartridge, 0,716 M, 1
CAS Nummer:  
Preis
64.60 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    EDTA digital titrator cartridge, 0,716 M