AuftragsverfolgungKontakt
Test kits, digital titration method | Avantor
Test kits, digital titration method
Chloride reagent kit: silver nitrate method, 10 - 10000 mg/l, 1 KIT
Katalognr. HACH2288000
Anbieter:   Hach
Test kits, digital titration method
Katalognr. HACH2288000
Anbieter:  Hach
CAS Nummer:  
Test kits, digital titration method
Test kits, digital titration method
Chloride reagent kit: silver nitrate method, 10 - 10000 mg/l, 1 KIT
CAS Nummer:  
Preis
342.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Achtung! Bitte beachten: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    Chloride reagent kit: silver nitrate method
  • Messbereich:
    10 - 10000 mg/l