Ampoule breakers
Ampoule breaker, Voluette ampoules
Katalognr. HACH2196800
Anbieter: Hach
Direkt zu
Product Details & Documents
These breakers for ampoules are designed to prevent contamination. Breaker snaps ampoule tip off.
Technische Daten
- Bezeichnung:Ampoule breaker
- Seq:03
- Für:Voluette ampoules