Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Membrane filters, Isopore™

TSTP, 3,00 µm, 47 mm
Catalog # MILFTSTP04700
Supplier:   Merck
Membrane filters, Isopore™
Catalog # MILFTSTP04700
Supplier:  Merck
Membrane filters, Isopore™
Membrane filters, Isopore™

TSTP, 3,00 µm, 47 mm

Price
423.00 SFr.
Log in for availability
1 * 100 ST
Product Details & Documents

These PC hydrophilic membrane are track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. They are composed of polycarbonate film, which has a smooth, glass like surface for clearer sample observation.

  • Membrane for microscopy and visual analysis
  • Membrane structure retains particles on the surface, simplifying counting and analysis
  • Do not stain, resulting in low background interference
  • Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
  • Can be sterilised by ethylene oxide, gamma irradiation or autoclave

The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. They are used for air monitoring, epifluorescent microscopy and chemotaxis assays.

Surface: plain

Operating temperature: 140 °C

Gravimetric extractables: <1,0%

Specifications
  • Ø:
    47 mm
  • Porengröße:
    3,00 µm
  • Typ:
    TSTP
  • Seq:
    34
  • Cat. No.:
    MILFTSTP04700
  • Luftdurchsatz:
    50 l/min/cm²
  • Stärke:
    22 µm
  • Wasserdurchfluss:
    180 ml/min/cm²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.