Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Dishes, evaporating | Avantor

Dishes, evaporating

250 ml, 45 mm, 85 mm
Katalognr. 216-2706
Anbieter:   Agencinox
Dishes, evaporating
Katalognr. 216-2706
Anbieter:  Agencinox
Dishes, evaporating
Dishes, evaporating

250 ml, 45 mm, 85 mm

Preis
SFr. 59.50
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Made of stainless steel.

  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
Technische Daten
  • Ext.Ø:
    85 mm
  • Kapazität:
    250 ml
  • Höhe:
    45 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.