Standard conical ground joints cones, long tip, Quickfit®
19/26 NS, 18,0 mm
201-8201
PYREX® borosilicate glass.
- Useful for construction of stirrer guides
- Avoids the problem of the badge charring at high temperatures which gives an untidy appearance
- Long tip cones manufactured with a minimum shank
Unbadged ground glass cones are free from ceramic markings.
Total length: 90 mm
- Kern:19/26 NS
- Rohr A-Ø:18,0 mm
- Seq:03