Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Standard conical ground joints cones, long tip, Quickfit®
19/26 NS, 18,0 mm
  201-8201
 :   SciLabware
Standard conical ground joints cones, long tip, Quickfit®
  201-8201
Standard conical ground joints cones, long tip, Quickfit®
Standard conical ground joints cones, long tip, Quickfit®
19/26 NS, 18,0 mm
SFr. 116.00
1 * 10 ST
PYREX® borosilicate glass.
  • Useful for construction of stirrer guides
  • Avoids the problem of the badge charring at high temperatures which gives an untidy appearance
  • Long tip cones manufactured with a minimum shank
Unbadged ground glass cones are free from ceramic markings.
Total length: 90 mm
  • Kern:
    19/26 NS
  • Rohr A-Ø:
    18,0 mm
  • Seq:
    03