Test Tubes with Rim, Medium Wall
180 mm, 18 mm
Katalognr. 212-0631
Anbieter: SciLabware
Direkt zu
Product Details & Documents
PYREX®, borosilicate glass.
- Superior performance in comparison to neutral borosilicate and soda-lime glass
- Excellent resistance to chemicals and high temperature
High quality test tubes designed and constructed for optimum performance.
Certifications: ISO 4142
Technische Daten
- Ext.Ø:18 mm
- Länge:180 mm
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



