Sample bottles for push-on caps
31 ml, 62,2 mm, 30 mm
Katalognr. 215-2627
Anbieter: GRAVIS
Direkt zu
Product Details & Documents
Chemistry glass, white or amber, without push-on cap.
- Amber glass to protect light-sensitive materials
Ordering information: Push-on PE caps are available separately.
Documents
SPVAE00943_TDS_EN
Datasheet
Technische Daten
- Kapazität:31 ml
- Höhe:62,2 mm
- Ø Hals:30 mm