Harnstoff
Glasflasche für Feststoffe, 250 g
Katalognr. NADIEC-605-250G
Marketsource
Anbieter: NATIONAL DIAGNOSTICS
Direkt zu
Product Details & Documents
Synonyme: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Nummer:57-13-6
- Formel:NH₂CONH₂
- MDL-Nummer:MFCD00008022
- Molekulargewicht:60.06 g/mol
- Merck-Index:13,09935
- Siedepunkt:197 °C (1013 hPa)
- Schmelzender Platingehalt:132,7 °C
- Dichte:1,34 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-315-5
- ERREICHEN:01-2119463277-33
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



