Spitzkolben, mit Normschliff
250 ml, 19/26 NS, 14/23 NS, 85 mm
Katalognr. LENZ03222149
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Technische Daten
- Ext.Ø:85 mm
- Kapazität:250 ml
- Seitenhälse:14/23 NS
- Mittelhals:19/26 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



