Spitzkolben, mit Normschliff
100 ml, 19/26 NS, 14/23 NS, 64 mm
Katalognr. LENZ03222137
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Technische Daten
- Ext.Ø:64 mm
- Kapazität:100 ml
- Seitenhälse:14/23 NS
- Mittelhals:19/26 NS



