Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Spitzkolben, mit Normschliff

Spitzkolben, mit Normschliff

200 ml, 14/23 NS, 14/23 NS, 85 mm
Catalog # LENZ03221149
Supplier:   Lenz Laborglas
Spitzkolben, mit Normschliff
Catalog # LENZ03221149
Supplier:  Lenz Laborglas
Spitzkolben, mit Normschliff
Spitzkolben, mit Normschliff

200 ml, 14/23 NS, 14/23 NS, 85 mm

Price
SFr. 117.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

The conical geometry makes them ideal for small-scale reactions.

Documents

Specifications
  • Ext.Ø:
    85 mm
  • Kapazität:
    200 ml
  • Seitenhälse:
    14/23 NS
  • Mittelhals:
    14/23 NS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.