Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Distilling receivers, Bernauer | Avantor
Distilling receivers, Bernauer
29/32 NS
Katalognr. LENZ05288004
Anbieter:   Lenz Laborglas
Distilling receivers, Bernauer
Katalognr. LENZ05288004
Anbieter:  Lenz Laborglas
Distilling receivers, Bernauer
Distilling receivers, Bernauer
29/32 NS
Preis
188.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Dokumente
Technische Daten
  • Hülse:
    29/32 NS