RIBOJUICE SMALL RNA TRANSFECTION REAGENT 1 * 0,3 mL
Catalog # 71115-3
Supplier: Merck Millipore (Novagen)
RIBOJUICE SMALL RNA TRANSFECTION REAGENT 1 * 0,3 mL
Catalog # 71115-3
Supplier: Merck Millipore (Novagen)
Frequently Bought Together
VWR® Acrylamid-Bis (29:1), 30 % wässrige Lösung, hochreine Qualität
Supplier: VWR Chemicals
DNA ladders, GeneRuler™ and O'GeneRuler™
Supplier: Thermo Fisher Scientific
Kits for Precellys®
Supplier: Bertin Instruments
VWR®, Kryoboxen, Karton, 133×133 mm
Supplier: VWR Collection
Dekontaminationslösung RNase AWAY®
Supplier: Thermo Fisher Scientific
[EN]MARKERS, 1BP + 1500BP, 3.2ML 1 * 3,2 mL
Supplier: AGILENT
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Ribonuclease A
Supplier: PanReac AppliChem
Restriction enzymes, BamHI, HC, Fermentas
Supplier: Thermo Fisher Scientific
[EN]FEMTO PULSE INTERCALATING DYE, 30UL 1 * 30 µG
Supplier: AGILENT
HUMAN GENOMIC DNA 1 * 100 µG
Supplier: Merck Millipore (Novagen)
DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT