Inlet liners
Splitless, single taper, direct connect liner, Agilent proprietary deactivation, 4 mm
Katalognr. HEWLG1544-80700
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Technische Daten
- Ø int.:4 mm
- Bezeichnung:Splitless, single taper, direct connect liner, Agilent proprietary deactivation
- Seq:50