AuftragsverfolgungKontakt
Preventive maintenance kit for LC/MS | Avantor
Preventive maintenance kit for LC/MS
Hydro moisture trap, refillable, 6,4 mm fittings, LC/MS
Katalognr. HEWLHMT200-4
Preventive maintenance kit for LC/MS
Katalognr. HEWLHMT200-4
Preventive maintenance kit for LC/MS
Preventive maintenance kit for LC/MS
Hydro moisture trap, refillable, 6,4 mm fittings, LC/MS
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Technische Daten
  • Für:
    LC/MS
  • Bezeichnung:
    Hydro moisture trap, refillable, 6,4 mm fittings
  • Seq:
    11