AuftragsverfolgungKontakt
Retort rods, unthreaded
Retort rods, unthreaded
13 mm, 1000 mm
Catalog # 241-2116
Supplier:   HARVARD APPARATUS
Retort rods, unthreaded
Catalog # 241-2116
Supplier:  HARVARD APPARATUS
Retort rods, unthreaded
13 mm, 1000 mm
Price
SFr. 81.10
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Brilliantly polished stainless steel with good surface hardness that will not score from clamp binding screws
  • Hollow, lightweight rods (Ø 13 mm) with continuously welded walls (1,6 mm thick)
  • Good chemical resistance
  • Red caps
Specifications
  • Ø:
    13 mm
  • Länge:
    1000 mm
  • Seq:
    06
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..