Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Trolleys, Variocar® 60 basic | Avantor

Trolleys, Variocar® 60 basic

Trolley, with one drawer, 600×450×730 mm
Katalognr. 139-9973
Trolleys, Variocar® 60 basic
Katalognr. 139-9973
Trolleys, Variocar® 60 basic
Trolleys, Variocar® 60 basic

Trolley, with one drawer, 600×450×730 mm

Preis
SFr. 1’260.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Multipurpose trolleys with chromium plated frame and shelves.

  • White shelves and drawers with designed drawer handles in different colours
  • Castors of 75 mm Ø each
  • Total loading capacity up to 50 kg
Technische Daten
  • Bezeichnung:
    Trolley, with one drawer
  • B×T×H:
    600×450×730 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.