AuftragsverfolgungKontakt
Brandschutzzeichen
Brandmeldetelefon, Leuchtfolie, 200×200 mm
  116-2026
 :   KROSCHKE
Brandschutzzeichen
  116-2026
 :  KROSCHKE
Brandschutzzeichen
Brandschutzzeichen
Brandmeldetelefon, Leuchtfolie, 200×200 mm
SFr. 264.00
1 * 1 ST
These self-adhesive, PVC film signs are used to indicate fire equipment. According to EC Directive 92/58/EEC and related national legislation on safety and/or health signs and also according to to VBG 125 (a regulation of the German employer's liability insurance association), signboards/signs for fire equipment must be permanently fitted and clearly recognisable at all times.
  • Suitable for use inside and outside
  • Visible from a distance of ∼12 metres
  • Long-lasting persistent luminescence material available
 : VBG 125 (now known as BGV A8).
 : Long-lasting persistent luminescence fire signs must be displayed in new and refurbished buildings if no safety lighting is available.
  • Bezeichnung:
    Brandmeldetelefon
  • Abmessungen:
    200×200 mm
  • Material:
    Leuchtfolie