pH-Kombinationselektroden
Sensolyt® ECA pH-Einstabmeßkette**, Sensolyt® ECA, zur Installation ins SensoLyt® Fitting, 2 - 12, Normal kontaminiertes Abwasser
Katalognr. 708-0057
Anbieter: WTW ONLINE
Direkt zu
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Technische Daten
- Für:Normal kontaminiertes Abwasser
- Typ:Sensolyt® ECA, zur Installation ins SensoLyt® Fitting
- Bezeichnung:Sensolyt® ECA pH-Einstabmeßkette**
- Seq:35
- pH:2 - 12