PROTEINASE K SOLUTION, 600MAU/ML 1 * 2 mL
71049-3
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
TRIS Pufferlösung pH 8 (1 mol/l), Millipore®
: Merck Millipore (Calbiochem)
DNA AWAY® Dekontaminationslösung
: Thermo Fisher Scientific
VWR®, Transferpipetten, Standard Line
: VWR Collection
VWR®, Transferpipetten, High Performance
: VWR Collection
VWR®, Mikrozentrifugenröhrchen, leistungsstark
: VWR Collection
VWR®, Petrischalen, PS
: VWR Collection
VWR®, PCR-Gefäße, 0,2 ml
: VWR Collection
Einwegspritzen, dreiteilig, HENKE-JECT®
: HENKE SASS WOLF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF