Basalmembranmatrix, Corning® Matrigel®
Product Details & Documents
Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation.
- Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
- Extensive PCR testing is performed to screen for a number of pathogens
- Tested and found negative for bacteria, fungi, and mycoplasma
- Protein concentrations determined by Lowry method
- Endotoxin units measured by Limulus Amoebocyte Lysate assay
- Matrigel Matrix gel stability tested for a period of 14 days at 37°C
- Biological activity determined for each lot using a neurite outgrowth assay.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws
Note: For research use only. Not intended for diagnostic or therapeutic procedures.
When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.
Dokumente
Zertifikate
Suche nach ChargennummerCellular Analysis of 3D Spheroid-Based Tumor Invasion Assays Corning® Matrigel® Basement Membrane, Matrix for 3D Culture In Vitro Corning® Matrigel® Matrix, Frequently Asked Questions Culture and Assay Systems Used for 3D Cell Culture Methods for Implantation of Corning® Matrigel® Matrix into Mice and Tissue Fixation
Corning® Matrigel® Matrix-3D Plates for High Throughput 3D Assays Culture of Mouse Intestinal Organoids in Corning® Matrigel® Matrix for Organoid Culture
Anwendungshinweise
sds_BDAA354234 sds_BDAA356230
Sicherheitsdatenblatt (SDS)
Technische Daten
Technische Daten
- VE:10 mL
- Bezeichnung:Corning® Matrigel® Basalmembranmatrix mit reduziertem Wachstumsfaktor
- Cat. No.:734-0269
- Ursprung:Engelbreth-Holm-Swarm mouse tumour
- Qualitätskontrolle:Auf die Fähigkeit getestet, schnell in Gelform überzugehen und diesen Zustand mit Nährmedien für eine Dauer von 14 Tagen bei 37 °C stabil zu halten. <br />Auf die Fähigkeit getestet, Neuritauswuchs von Ganglienzellen der Dorsalwurzel (Küken) zu fördern. <br />Auf Bakterien, Pilze und Mycoplasma negativ getestet. <br />Auf Endotoxin getestet (LAL-Assay)
- Lagerung and Stabilität:Bei -20 °C mindestens 3 Monate oder bei 37 °C 12 Tage stabil
- Umweltzertifizierung:ISO 14001 - Bedford
- Umweltfreundlichkeit:Umweltschonende Herstellung
Technische Daten
Complementary Products
Produktinformationen
- Katalognr.ModellAuflösungBezeichnungVerpacktMembranPorengrößeKulturflächeEmpf. ArbeitsvolumenMembran-ØFarbeVerfügbarkeitPreis
- MLDVMICRO4ImageXpress Micro 425 - 100 nmHigh-Content-Imaging-SystemEinloggen für Verfügbarkeit1 * 1 ST - SFr. 0.00
- 735-0364MIKROKONFOKAL25 nmHigh-content mikrokonfokales BildgebungssystemEinloggen für Verfügbarkeit1 * 1 ST - SFr. 0.00
- MLDVNANOMXImage Xpress NanoImaging-System mit MetaXpress-Software001Einloggen für Verfügbarkeit1 * 1 ST - SFr. 0.00
- 734-27156 Einsätze/Platte, 4 Platten/KartonTransluzent0,1 µm2,6 ml6-Well-Platte mit Einsätzen0124 mmEinloggen für Verfügbarkeit1 * 24 ST - SFr. 196.00
- 734-2763Schwarz/klar200 µl+Half area microplate, with lid, tissue culture treated01Einloggen für Verfügbarkeit1 * 16 ST - SFr. 443.00
- 392-0608Klar400 µl+MicroWell™ 96-Well-Platte, flacher Boden, Nunclon™ Sphera™ behandelte Oberfläche06Einloggen für Verfügbarkeit1 * 8 ST - SFr. 221.00
- 738-0204Schwarz/klar300 µl+Round bottom, spheroid with lid, ultra-low attachment surface01Einloggen für Verfügbarkeit1 * 5 ST - SFr. 263.00
- 734-2971Einzeln verpackt113 cm²Standard-12-Well-Platte, 6× oberflächenbehandelte Scaffold-Einsätze (21,0×1,6 mm)Einloggen für Verfügbarkeit1 * 8 ST - SFr. 273.00
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



