Cold traps with/without condensation drain
200 ml, without condensate drain
Katalognr. 181-9982
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump.
DURAN®, klar.
- Sehr gute chemische Beständigkeit
- Hohe Temperaturbeständigkeit
- Minimale Wärmeausdehnung, dadurch bedingt hohe Temperaturwechselbeständigkeit
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Technische Daten
- Bezeichnung:200 ml, without condensate drain
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.