Spitzkolben, mit Normschliff
100 ml, 14/23 NS, 14/23 NS, 64 mm
271-1234
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
- Ext.Ø:64 mm
- Kapazität:100 ml
- Seitenhälse:14/23 NS
- Mittelhals:14/23 NS



