Exsikkator-Platten
Porzellan, 140 mm
Katalognr. BRND65965
Anbieter: Brand
Direkt zu
Product Details & Documents
Desiccator plates for glass and plastic desiccators.
- Central hole measures approximately 20 mm diameter
- Small perforations measure approximately 5 mm diameter
Technische Daten
- Ø:140 mm
- Typ:Porzellan
- Seq:01