Kegelschliffstopfen
14/23, Klarglas, massiv
Katalognr. BRND144520
Anbieter: Brand
Direkt zu
Product Details & Documents
Boro 3.3.
- Massive bzw. halbhohle Ausfühung, mit Achtkantgriffscheibe
Certifications: DIN 12252
Technische Daten
- NS:14/23
- Typ:Klarglas, massiv
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.