n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
Glasflasche, 1 L
Anbieter: Avantor
n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
Katalognr. 9304.1000
Anbieter: Avantor
CAS Nummer: 110-54-3
Direkt zu
Product Details & Documents
for use in High Performance Liquid Chromatography
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- UN:1208
- ADR:3,II
Technische Daten
Ergebnisse des Spezifikationstests
- Assay (by GC):min. 95 %
- Residue after Evaporation:max. 2 ppm
- Substances Darkened by H2SO4:passes test
- Water (H2O):max. 0,01 %
- Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).:-
- at 210 nm:max. 0,2
- at 220 nm:max. 0,08
- at 254 nm:max. 0,008
- at 280 nm:max. 0,005
- at 350 nm:max. 0,005
- UV Cut-off, nm:max. 192
Technische Daten
- Packung:Glasflasche
- VE:1 L



