Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Verschlüsse 11 mm, WHEATON® | Avantor
Verschlüsse 11 mm, WHEATON®
Natural PE, with cross slit, 11 mm Hals, PTFE/silicone
Katalognr. 548-2114
Anbieter:   DWK Life Sciences
Verschlüsse 11 mm, WHEATON®
Katalognr. 548-2114
Anbieter:  DWK Life Sciences
Verschlüsse 11 mm, WHEATON®
Verschlüsse 11 mm, WHEATON®
Natural PE, with cross slit, 11 mm Hals, PTFE/silicone
Preis
SFr. 1’420.00
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents
Technische Daten
  • Für:
    11 mm Hals
  • Septum:
    PTFE/silicone
  • Typ:
    Natural PE, with cross slit
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.