Jars, with lid
50 mm, 30 mm
Katalognr. 525-1364
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Technische Daten
- Ext.Ø:50 mm
- Höhe:30 mm