Wolframdichloriddioxid ≥99%
Glasflasche für Feststoffe, 50 g
Anbieter: Thermo Fisher Scientific
Wolframdichloriddioxid ≥99%
Katalognr. 89962.18
Anbieter: Thermo Fisher Scientific
CAS Nummer: 13520-76-8
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 13520-76-8 Molecular Formula: WO2Cl2 Formula Weight: 286.75 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3260 MDL No.: MFCD00054136
- CAS Nummer:13520-76-8
- Formel:Cl₂O₂W
- MDL-Nummer:MFCD00054136
- Molekulargewicht:286.75 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:236-862-1
- ADR:8,III
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:50 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



