Silberchlorid ≥95%
100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 95% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399
Silver plating
- CAS Nummer:7783-90-6
- Formel:AgCl
- MDL-Nummer:MFCD00003399
- Molekulargewicht:143.32 g/mol
- Merck-Index:14,8509
- Siedepunkt:1550 °C
- Schmelzender Platingehalt:455 °C
- Dichte:5,56
- Lagertemperatur:Raumtemperatur
- EINECS:232-033-3
- ADR:9,III
Dokumente
Technische Daten
Technische Daten
- VE:100 g



